Order Entry
Denmark
ContactUsLinkComponent
Clostridium sporogenes WDCM 00008 Vitroids™, Certified Reference Material, Suitable for Microbiology, Supelco®
Clostridium sporogenes WDCM 00008 Vitroids™, Certified Reference Material, Suitable for Microbiology, Supelco®
Catalog # VT000082-10EA
Supplier:  Merck
CAS Number:  
Clostridium sporogenes WDCM 00008 Vitroids™, Certified Reference Material, Suitable for Microbiology, Supelco®
Catalog # VT000082-10EA
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    10
  • Description:
    Clostridium sporogenes WDCM 00008 Vitroids™,15 - 80 CFU mean value range
  • M-Clarity quality level:
    MQ300
  • Storage temperature:
    –20 °C
  • Product type:
    Hydrosoluble discs

Specifications

About this item

Derived from CECT® strains.

Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025.