Citrobacter freundii CECT® 401 Vitroids™, Certified Reference Material, Suitable for Microbiology, Supelco®
VT004016-10EA
: Merck
:
- Pk:10
- Description:Citrobacter freundii CECT® 401 Vitroids™, 1000 - 10,000 CFU mean value range
- M-Clarity quality level:MQ300
- Storage temperature:–20 °C
- Product type:Hydrosoluble discs
Derived from CECT® strains.
: This certified reference material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025.