Candida albicans NCPF 3255 Lenticule® Discs, Suitable for Microbiology, Reference Material, Supelco®
Supplier: Merck
Certificates
About this item
Certifications: This reference material (RM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025.
Caution: LENTICULE® discs are manufactured by Sigma-Aldrich under licence from Public Health England.
Specifications
- M-Clarity quality level:MQ200
- Storage temperature:−20 °C
- Product type:Hydrosoluble discs