About this item
These Vitroids™ discs contain Candida albicans WDCM 00054 viable microorganisms in a certified quantity (generally accredited according to ISO/IEC 17025), produced under reproducible conditions compliant with ISO 17034:2016 using authenticated strains from CECT®.
Applications:
- Daily quality control
- Performance testing of media acc. ISO 11133
- Validation of new methods
- Materials for proficiency testing or ring trials
- Method development
- Staff training
- Starter cultures
The discs consist of pure cultures of bacteria in a solid matrix, stable for at least one year and are in a viable state with a shelf life of 1-3 years.
Each batch is provided with a comprehensive certificate of analysis that specifies the mean number of colony-forming units (CFU), an expanded uncertainty about the mean, details about the method used to determine the product data and the number of passages (subcultures) from the original strain.
Vitroids™ are derived from a traceable culture obtained freeze-dried from CECT®.
Specifications
- M-Clarity quality level:MQ300
- Storage temperature:−20 °C
- Product type:Hydrosoluble discs