Order Entry
Denmark
ContactUsLinkComponent
Aspergillus brasiliensis (formerly Aspergillus niger) NCPF 2275 LENTICULE® Discs, Suitable for Microbiology, Reference Material
Aspergillus brasiliensis (formerly Aspergillus niger) NCPF 2275 LENTICULE® Discs, Suitable for Microbiology, Reference Material
Catalog # RMF02275L-10EA
Supplier:  Merck
CAS Number:  
Aspergillus brasiliensis (formerly Aspergillus niger) NCPF 2275 LENTICULE® Discs, Suitable for Microbiology, Reference Material
Catalog # RMF02275L-10EA
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    10
  • Description:
    Aspergillus brasiliensis NCPF 2275 LENTICULE® discs, 30-120 CFU
  • M-Clarity quality level:
    MQ200
  • Storage temperature:
    −20 °C
  • Product type:
    Hydrosoluble discs

Specifications

About this item

Certifications: This reference material (RM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025.
Caution: Lenticule is a registered trademark of United Kingdom Secretary of State for Health