Anaerobic systems, workstation, Whitley A95
Katalog # DOWIA07400
Leverandør: DON WHITLEY SCIENTIFIC
Specifikationer
- Kapacitet:Working: 960 plates;
Incubation: 1120 plates;
Port/air lock: 10 plates p/port + 90 plates via airlock - Beskrivelse:A95 Anaerobic workstation
- Environmentally Preferable:
- Adgangsporte:Manual
- Cyklustid:5 min (airlock)
- Gas type:ANO2 / N2
- Pakn:1
Specifikationer
Om denne vare
The A95 Workstation has a huge capacity of up to 1120×90mm Petri dishes, depending upon the way you choose to configure your working and incubation areas.
- Innovative touch screen interface, which allows the incorporation of some unique features and benefits
- The four manually operated, oval glove ports enable two people to operate within the same controlled atmosphere simultaneously
- Rapid, built-in airlock with mechanical and electrical interlocks -no risk of compromising conditions inside the chamber, flushed with nitrogen as an additional cost saving
- Optional anaerobic conditions monitor
- Dual gas operation for real savings on gas
- Automatic commissioning cycle
- Automatic de-humidification system – no user maintenance necessary
- Optional Three Gas (TG) version available to help reduce running costs and provide additional flexibility
This model is ideal for tutoring or for very busy laboratories. As well as having a huge 36 litre airlock, each port also acts as a mini airlock so you can transfer 10 plates when you insert or withdraw each arm, reducing gas usage and the number of airlock cycles required.
Note: The cycle time mentioned in the specifications is for the airlock.
Ordering information: Options and accessories not included (sold separately). For more information on the available options and accessories, please see the attached brochures and contact your Avantor representative.