Order Entry
Denmark
ContactUsLinkComponent
Anaerobic systems, workstation, Whitley A55
Anaerobic systems, workstation, Whitley A55
Catalog # DOWIA07760
Anaerobic systems, workstation, Whitley A55
Catalog # DOWIA07760

Specifications

  • Capacity:
    Working: 1120 plates;
    Incubation: 1240 plates;
    Port/air lock: 40 plates via each airlock (2 airlocks)
  • Description:
    A55 Anaerobic workstation
  • Environmentally Preferable:
  • Access ports:
    Manual ports or instant access
  • Cycle time:
    60 s (airlock)
  • Gas type:
    ANO2 / N2
  • Pk:
    1

Specifications

About this item

The Whitley A55 has a chamber size twice the width of an A35, with a huge working and incubation area and airlocks at both ends.

  • The choice of conventional oval sleeved ports or a mixture of oval and Instant Access Ports
  • 2x rapid, built-in airlocks; flushed with nitrogen as an additional cost saving
  • Innovative colour touch screen interface with PIN-code protected user levels
  • Dual gas operation for real savings on gas
  • Comfortable to work in and excellent visibility
  • Optional anaerobic conditions monitor
  • Available as a dual temperature version (optional) allowing different temperatures in each half
  • Also available with the Whitley Internal HEPA Filtration System (optional)

Petri dishes and samples can be introduced and removed through either airlock creating a natural, Lean rhythm for your laboratory workflow. The A55 is ideal for the busy lab where organisation of samples and cultures is key as it can accommodate up to 1200 x 90mm Petri dishes in racks.

Note: The cycle time mentioned in the specifications is for the airlock.

Ordering information: Options and accessories not included (sold separately). For more information on the available options and accessories, please see the attached brochures and contact your Avantor representative.