Order Entry
Denmark
Orders LinkContactUsLinkComponent
Rectangle Cryogenic Labels for Thermal Transfer Barcode Printers
Supplier:   GA
Rectangle Cryogenic Labels for Thermal Transfer Barcode Printers
Supplier:  GA
Certificates
Rectangle Cryogenic Labels for Thermal Transfer Barcode Printers
Rectangle Cryogenic Labels for Thermal Transfer Barcode Printers
109 Options Available Below
Product Details & Documents
These labels are available in a range of sizes and configurations, perfect for identifying small diameter cryo vials, microtubes, cardboard or polypropylene cryo/freezer boxes, cell culture plates, and plastic bags.
  • Temperature range: −196 to +110 °C (−321 to +230 °F)
  • Resistant to long term cryogenic storage in freezers and liquid nitrogen (−196 °C/−321 °F)
  • Resistant to spraying and swabbing with alcohol and surface sanitising wipes
  • Require no cure time before liquid nitrogen immersion; will also adhere to wet surfaces
  • High-quality thermal transfer printing, ideal for serialised data and barcodes
  • Available in a large range of sizes, colours, and configurations
  • RR-class resin ribbon recommended
These thermal transfer cryogenic labels have a strong permanent adhesive that will remain firmly and permanently attached in ultra-low temperatures. They are designed for long-term cryo-preservation under cryogenic laboratory conditions, including in liquid and vapour phase liquid nitrogen (−196 °C/−321 °F), lab freezers (−80 °C/−112 °F), and dry ice transportation. JTTA labels require zero cure time and can be immersed in liquid nitrogen immediately after application without the need for additional tape or lamination. Additionally, these labels are waterproof, weatherproof, and durable. JTTA labels will adhere to wet surfaces, resist multiple freeze-thaw cycles, and can withstand elevated temperatures up to 110 °C.
They are also resistant to spraying with ethanol, isopropanol, and other alcohols, bleach, and detergents (testing recommended), as well as routine wiping and rubbing with surface sanitizing wipes such as Sani- Wipe™, Eco-Wipe™, and Eco-Wipe™ Duo.
These labels can be printed with all major brands of thermal-transfer printers. They are available in a range of colours and sizes and can be printed with alphanumeric text, as well as 1D and 2D barcodes.
Product Family Options
Product Information
  • Colour
    Dimensions
    Packed
    Availability
    Price
  • GAINJTTA-204C1-2WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    White
    11,9×11,9 mm (0,47×0,47")
    2000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 577,00 kr
    577,00 kr
    1 * 1 ROL
       577,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-204C1-2WH
  • GAINJTTA-204C3-4WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    White
    11,9×11,9 mm (0,47×0,47")
    4000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 1.160,00 kr
    1.160,00 kr
    1 * 1 ROL
       1.160,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-204C3-4WH
  • GAINJTTA-162C1-2WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    White
    12,7×25,4 mm (0,5×1")
    2000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 721,00 kr
    721,00 kr
    1 * 1 ROL
       721,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-162C1-2WH
  • GAINJTTA-277C1-1WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    White
    15,9×30,5 mm (0,63×1,2")
    1000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 356,00 kr
    356,00 kr
    1 * 1 ROL
       356,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-277C1-1WH
  • GAINJTTA-277C3-3WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    White
    15,9×30,5 mm (0,63×1,2")
    3000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 1.070,00 kr
    1.070,00 kr
    1 * 1 ROL
       1.070,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-277C3-3WH
  • GAINJTTA51NPC1-2WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    White
    19,1×10,2 mm (0,75×0,4")
    2000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 721,00 kr
    721,00 kr
    1 * 1 ROL
       721,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA51NPC1-2WH
  • GAINJTTA-49C1-2WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    White
    19,1×15,9 mm (0,75×0,63")
    2000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 697,00 kr
    697,00 kr
    1 * 1 ROL
       697,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-49C1-2WH
  • GAINJTTA-49C3-2WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    White
    19,1×15,9 mm (0,75×0,63")
    2000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 697,00 kr
    697,00 kr
    1 * 1 ROL
       697,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-49C3-2WH
  • GAINJTTA-69C1-2WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    White
    19,1×19,1 mm (0,75×0,75")
    2000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 806,00 kr
    806,00 kr
    1 * 1 ROL
       806,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-69C1-2WH
  • GAINJTTA-69C3-2WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    White
    19,1×19,1 mm (0,75×0,75")
    2000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 806,00 kr
    806,00 kr
    1 * 1 ROL
       806,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-69C3-2WH
  • GAINJTTA-177C1-1WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    White
    19,1×38,1 mm (0,75×1,5")
    1000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 521,00 kr
    521,00 kr
    1 * 1 ROL
       521,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-177C1-1WH
  • GAINJTTA213NPC15WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    White
    19,1×5,1 mm (0,75×0,2")
    5000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 1.450,00 kr
    1.450,00 kr
    1 * 1 ROL
       1.450,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA213NPC15WH
  • GAINJTTA-98C1-1WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    White
    22,2×22,2 mm (0,87×0,87")
    1000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 330,00 kr
    330,00 kr
    1 * 1 ROL
       330,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-98C1-1WH
  • GAINJTTA-98C3-3WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    White
    22,2×22,2 mm (0,87×0,87")
    3000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 989,00 kr
    989,00 kr
    1 * 1 ROL
       989,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-98C3-3WH
  • GAINJTTA188NPC13WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    White
    22,9×12,7 mm (0,9×0,5")
    3000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 865,00 kr
    865,00 kr
    1 * 1 ROL
       865,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA188NPC13WH
  • GAINJTTA-117C1-3WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    White
    22×15 mm (0,87×0,59")
    3000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 865,00 kr
    865,00 kr
    1 * 1 ROL
       865,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-117C1-3WH
  • GAINJTTA-117C3-4WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    White
    22×15 mm (0,87×0,59")
    4000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 1.140,00 kr
    1.140,00 kr
    1 * 1 ROL
       1.140,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-117C3-4WH
  • GAINJTTA-67C1-1WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    White
    23,8×23,8 mm (0,94×0,94")
    1000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 349,00 kr
    349,00 kr
    1 * 1 ROL
       349,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-67C1-1WH
  • GAINJTTA-67C3-1WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    White
    23,8×23,8 mm (0,94×0,94")
    1000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 349,00 kr
    349,00 kr
    1 * 1 ROL
       349,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-67C3-1WH
  • GAINJTTA-152C1-1WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    White
    23×19,1 mm (0,9×0,75")
    1000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 324,00 kr
    324,00 kr
    1 * 1 ROL
       324,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-152C1-1WH
  • GAINJTTA-152C3-6WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    White
    23×19,1 mm (0,9×0,75")
    6000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 1.940,00 kr
    1.940,00 kr
    1 * 1 ROL
       1.940,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-152C3-6WH
  • GAINJTTA-1C1-1WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    White
    25,4 × 11,1 mm (1×0,44")
    1000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 289,00 kr
    289,00 kr
    1 * 1 ROL
       289,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-1C1-1WH
  • GAINJTTA-1C3-6WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    White
    25,4 × 11,1 mm (1×0,44")
    6000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 1.730,00 kr
    1.730,00 kr
    1 * 1 ROL
       1.730,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-1C3-6WH
  • GAINJTTA-7C1-1WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    White
    25,4×12,7 mm (1×0,5")
    1000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 289,00 kr
    289,00 kr
    1 * 1 ROL
       289,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-7C1-1WH
  • GAINJTTA-7SBC1-1WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    White
    25,4×12,7 mm (1×0,5")
    1000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 289,00 kr
    289,00 kr
    1 * 1 ROL
       289,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-7SBC1-1WH
  • GAINJTTA-7C3-5WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    White
    25,4×12,7 mm (1×0,5")
    5000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 1.450,00 kr
    1.450,00 kr
    1 * 1 ROL
       1.450,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-7C3-5WH
  • GAINJTTA-4C1-1WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    White
    25,4×19,1 mm (1×0,75")
    1000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 333,00 kr
    333,00 kr
    1 * 1 ROL
       333,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-4C1-1WH
  • GAINJTTA-4C3-1WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    White
    25,4×19,1 mm (1×0,75")
    1000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 333,00 kr
    333,00 kr
    1 * 1 ROL
       333,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-4C3-1WH
  • GAINJTTA-29C1-1WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    White
    25,4×25,4 mm (1×1")
    1000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 361,00 kr
    361,00 kr
    1 * 1 ROL
       361,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-29C1-1WH
  • GAINJTTA-29C3-3WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    White
    25,4×25,4 mm (1×1")
    3000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 1.090,00 kr
    1.090,00 kr
    1 * 1 ROL
       1.090,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-29C3-3WH
  • GAINJTTA-118C1-1WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    White
    25,4×31,8 mm (1×1,25")
    1000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 442,00 kr
    442,00 kr
    1 * 1 ROL
       442,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-118C1-1WH
  • GAINJTTA-118C3-4WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    White
    25,4×31,8 mm (1×1,25")
    4000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 1.770,00 kr
    1.770,00 kr
    1 * 1 ROL
       1.770,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-118C3-4WH
  • GAINJTTA-94C1-1WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    White
    25,4×50,8 mm (1×2")
    1000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 721,00 kr
    721,00 kr
    1 * 1 ROL
       721,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-94C1-1WH
  • GAINJTTA209C3-10WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    White
    25,4×6,4 mm (1×0,25")
    10000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 2.840,00 kr
    2.840,00 kr
    1 * 1 ROL
       2.840,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA209C3-10WH
  • GAINJTTA-209C1-4WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    White
    25,4×6,4 mm (1×0,25")
    4000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 1.140,00 kr
    1.140,00 kr
    1 * 1 ROL
       1.140,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-209C1-4WH
  • GAINJTTA-225C1-3WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    White
    25,4×9,5 mm (1×0,375")
    3000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 865,00 kr
    865,00 kr
    1 * 1 ROL
       865,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-225C1-3WH
  • GAINJTTA-225C3-6WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    White
    25,4×9,5 mm (1×0,375")
    6000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 1.730,00 kr
    1.730,00 kr
    1 * 1 ROL
       1.730,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-225C3-6WH
  • GAINJTTA-57C1-1WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    White
    28,6×19,1 mm (1,12×0,75")
    1000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 349,00 kr
    349,00 kr
    1 * 1 ROL
       349,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-57C1-1WH
  • GAINJTTA-57C3-1WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    White
    28,6×19,1 mm (1,12×0,75")
    1000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 349,00 kr
    349,00 kr
    1 * 1 ROL
       349,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-57C3-1WH
  • GAINJTTA-57C3-1GA
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    Green
    28,6×19,1 mm (1,12×0,75")
    1000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 349,00 kr
    349,00 kr
    1 * 1 ROL
       349,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-57C3-1GA
  • GAINJTTA-18C1-2WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    White
    31,8×12,7 mm (1,25×0,5")
    2000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 704,00 kr
    704,00 kr
    1 * 1 ROL
       704,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-18C1-2WH
  • GAINJTTA-18C3-2WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    White
    31,8×12,7 mm (1,25×0,5")
    2000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 704,00 kr
    704,00 kr
    1 * 1 ROL
       704,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-18C3-2WH
  • GAINJTTA-9C1-1WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    White
    31,8×22,2 mm (1,25×0,875")
    1000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 394,00 kr
    394,00 kr
    1 * 1 ROL
       394,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-9C1-1WH
  • GAINJTTA-9C3-1WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    White
    31,8×22,2 mm (1,25×0,875")
    1000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 394,00 kr
    394,00 kr
    1 * 1 ROL
       394,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-9C3-1WH
  • GAINJTTA-58C1-1WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    White
    31,8×25,4 mm (1,25×1")
    1000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 442,00 kr
    442,00 kr
    1 * 1 ROL
       442,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-58C1-1WH
  • GAINJTTA-58C3-4WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    White
    31,8×25,4 mm (1,25×1")
    4000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 1.770,00 kr
    1.770,00 kr
    1 * 1 ROL
       1.770,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-58C3-4WH
  • GAINJTTA-65C1-1WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    White
    31,8×31,8 mm (1,25×1,25")
    1000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 539,00 kr
    539,00 kr
    1 * 1 ROL
       539,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-65C1-1WH
  • GAINJTTA-65C3-1WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    White
    31,8×31,8 mm (1,25×1,25")
    1000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 539,00 kr
    539,00 kr
    1 * 1 ROL
       539,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-65C3-1WH
  • GAINJTTA-169C1-4WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    White
    31,8×6,4 mm (1,25×0,25")
    4000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 1.140,00 kr
    1.140,00 kr
    1 * 1 ROL
       1.140,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-169C1-4WH
  • GAINJTTA-97C1-1WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    White
    32,5×15,9 mm (1,28×0,63")
    1000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 339,00 kr
    339,00 kr
    1 * 1 ROL
       339,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-97C1-1WH
  • GAINJTTA-97C3-2WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    White
    32,5×15,9 mm (1,28×0,63")
    2000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 675,00 kr
    675,00 kr
    1 * 1 ROL
       675,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-97C3-2WH
  • GAINJTTA-19C1-1WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    White
    33×15,9 mm (1,3×0,63")
    1000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 339,00 kr
    339,00 kr
    1 * 1 ROL
       339,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-19C1-1WH
  • GAINJTTA-19C3-2WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    White
    33×15,9 mm (1,3×0,63")
    2000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 675,00 kr
    675,00 kr
    1 * 1 ROL
       675,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-19C3-2WH
  • GAINJTTA-6C1-3WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    White
    34,9×12,7 mm (1,38×0,5")
    3000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 943,00 kr
    943,00 kr
    1 * 1 ROL
       943,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-6C1-3WH
  • GAINJTTA-168C1-2WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    White
    35,6×12,7 mm (1,4×0,5")
    2000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 673,00 kr
    673,00 kr
    1 * 1 ROL
       673,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-168C1-2WH
  • GAINJTTA-168C3-4WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    White
    35,6×12,7 mm (1,4×0,5")
    4000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 1.350,00 kr
    1.350,00 kr
    1 * 1 ROL
       1.350,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-168C3-4WH
  • GAINJTTA-536C1-2WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    White
    38,1×10,2 mm (1,5×0,4")
    2000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 655,00 kr
    655,00 kr
    1 * 1 ROL
       655,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-536C1-2WH
  • GAINJTTA-240C1-1WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    White
    38,1×12,7 mm (1,5×0,5")
    1000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 341,00 kr
    341,00 kr
    1 * 1 ROL
       341,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-240C1-1WH
  • GAINJTTA-240C3-3WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    White
    38,1×12,7 mm (1,5×0,5")
    3000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 1.020,00 kr
    1.020,00 kr
    1 * 1 ROL
       1.020,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-240C3-3WH
  • GAINJTTA-10C1-1WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    White
    38,1×19,1 mm (1,5×0,75")
    1000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 406,00 kr
    406,00 kr
    1 * 1 ROL
       406,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-10C1-1WH
  • GAINJTTA-10C3-2WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    White
    38,1×19,1 mm (1,5×0,75")
    2000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 810,00 kr
    810,00 kr
    1 * 1 ROL
       810,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-10C3-2WH
  • GAINJTTA-107C1-1WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    White
    38,1×22,2 mm (1,5×0,875")
    1000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 463,00 kr
    463,00 kr
    1 * 1 ROL
       463,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-107C1-1WH
  • GAINJTTA-107C3-3WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    White
    38,1×22,2 mm (1,5×0,875")
    3000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 1.390,00 kr
    1.390,00 kr
    1 * 1 ROL
       1.390,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-107C3-3WH
  • GAINJTTA-504C1-1WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    White
    38,1×25,4 mm (1,5×1")
    1000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 545,00 kr
    545,00 kr
    1 * 1 ROL
       545,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-504C1-1WH
  • GAINJTTA-504C3-3WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    White
    38,1×25,4 mm (1,5×1")
    3000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 1.630,00 kr
    1.630,00 kr
    1 * 1 ROL
       1.630,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-504C3-3WH
  • GAINJTTA-272C1-1WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    White
    38,1×31,8 mm (1,5×1,25")
    1000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 667,00 kr
    667,00 kr
    1 * 1 ROL
       667,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-272C1-1WH
  • GAINJTTA-272C3-4WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    White
    38,1×31,8 mm (1,5×1,25")
    4000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 2.670,00 kr
    2.670,00 kr
    1 * 1 ROL
       2.670,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-272C3-4WH
  • GAINJTTA-66C1-1WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    White
    38,1×38,1 mm (1,5×1,5")
    1000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 751,00 kr
    751,00 kr
    1 * 1 ROL
       751,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-66C1-1WH
  • GAINJTTA-66C3-2WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    White
    38,1×38,1 mm (1,5×1,5")
    2000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 1.510,00 kr
    1.510,00 kr
    1 * 1 ROL
       1.510,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-66C3-2WH
  • GAINJTTA-232C1-2WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    White
    41,3×15,9 mm (1,63×0,63")
    2000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 749,00 kr
    749,00 kr
    1 * 1 ROL
       749,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-232C1-2WH
  • GAINJTTA-232C3-4WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    White
    41,3×15,9 mm (1,63×0,63")
    4000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 1.500,00 kr
    1.500,00 kr
    1 * 1 ROL
       1.500,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-232C3-4WH
  • GAINJTTA-38C1-2WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    White
    44,5×12,7 mm (1,75×0,5")
    2000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 670,00 kr
    670,00 kr
    1 * 1 ROL
       670,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-38C1-2WH
  • GAINJTTA-38C3-6WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    White
    44,5×12,7 mm (1,75×0,5")
    6000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 2.010,00 kr
    2.010,00 kr
    1 * 1 ROL
       2.010,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-38C3-6WH
  • GAINJTTA-242C1-1WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    White
    44,5×19,1 mm (1,75×0,75")
    1000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 419,00 kr
    419,00 kr
    1 * 1 ROL
       419,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-242C1-1WH
  • GAINJTTA-242C3-3WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    White
    44,5×19,1 mm (1,75×0,75")
    3000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 1.260,00 kr
    1.260,00 kr
    1 * 1 ROL
       1.260,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-242C3-3WH
  • GAINJTTA-24C1-2WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    White
    44,5×25,4 mm (1,75×1")
    2000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 937,00 kr
    937,00 kr
    1 * 1 ROL
       937,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-24C1-2WH
  • GAINJTTA-24C3-2WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    White
    44,5×25,4 mm (1,75×1")
    2000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 937,00 kr
    937,00 kr
    1 * 1 ROL
       937,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-24C3-2WH
  • GAINJTTA-37C1-1WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    White
    48×31,8 mm (1,89×1,25")
    1000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 790,00 kr
    790,00 kr
    1 * 1 ROL
       790,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-37C1-1WH
  • GAINJTTA-128C1-1WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    White
    50,8×19,1 mm (2×0,75")
    1000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 481,00 kr
    481,00 kr
    1 * 1 ROL
       481,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-128C1-1WH
  • GAINJTTA-128C3-2WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    White
    50,8×19,1 mm (2×0,75")
    2000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 961,00 kr
    961,00 kr
    1 * 1 ROL
       961,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-128C3-2WH
  • GAINJTTA-28C1-2WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    White
    50,8×25,4 mm (2×1")
    2000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 1.140,00 kr
    1.140,00 kr
    1 * 1 ROL
       1.140,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-28C1-2WH
  • GAINJTTA-28C3-2WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    White
    50,8×25,4 mm (2×1")
    2000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 1.140,00 kr
    1.140,00 kr
    1 * 1 ROL
       1.140,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-28C3-2WH
  • GAINJTTA-14C1-1WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    White
    50,8×38,1 mm (2×1,5")
    1000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 984,00 kr
    984,00 kr
    1 * 1 ROL
       984,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-14C1-1WH
  • GAINJTTA-14C3-2WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    White
    50,8×38,1 mm (2×1,5")
    2000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 1.970,00 kr
    1.970,00 kr
    1 * 1 ROL
       1.970,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-14C3-2WH
  • GAINJTTA-27C1-1WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    White
    50,8×50,8 mm (2×2")
    1000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 1.140,00 kr
    1.140,00 kr
    1 * 1 ROL
       1.140,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-27C1-1WH
  • GAINJTTA-27C3-1WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    White
    50,8×50,8 mm (2×2")
    1000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 1.140,00 kr
    1.140,00 kr
    1 * 1 ROL
       1.140,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-27C3-1WH
  • GAINJTTA60NP10.5WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    White
    50,8×76,2 mm (2×3")
    500 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 641,00 kr
    641,00 kr
    1 * 1 ROL
       641,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA60NP10.5WH
  • GAINJTTA-206C1-2WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    White
    54×6,9 mm (2,12×0,27")
    2000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 601,00 kr
    601,00 kr
    1 * 1 ROL
       601,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-206C1-2WH
  • GAINJTTA-231C1-1WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    White
    57,2×25,4 mm (2,25×1")
    1000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 800,00 kr
    800,00 kr
    1 * 1 ROL
       800,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-231C1-1WH
  • GAINJTTA-32C1-1WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    White
    63,5×12,7 mm (2,5×0,5")
    1000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 450,00 kr
    450,00 kr
    1 * 1 ROL
       450,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-32C1-1WH
  • GAINJTTA-32C3-3WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    White
    63,5×12,7 mm (2,5×0,5")
    3000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 1.350,00 kr
    1.350,00 kr
    1 * 1 ROL
       1.350,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-32C3-3WH
  • GAINJTTA-68C1-2WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    White
    63,5×19,1 mm (2,5×0,75")
    2000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 1.290,00 kr
    1.290,00 kr
    1 * 1 ROL
       1.290,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-68C1-2WH
  • GAINJTTA-68C3-2WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    White
    63,5×19,1 mm (2,5×0,75")
    2000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 1.290,00 kr
    1.290,00 kr
    1 * 1 ROL
       1.290,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-68C3-2WH
  • GAINJTTA-5C1-1WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    White
    63,5×25,4 mm (2,5×1")
    1000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 840,00 kr
    840,00 kr
    1 * 1 ROL
       840,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-5C1-1WH
  • GAINJTTA-5C3-4WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    White
    63,5×25,4 mm (2,5×1")
    4000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 3.360,00 kr
    3.360,00 kr
    1 * 1 ROL
       3.360,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-5C3-4WH
  • GAINJTTA-238C1-1WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    White
    63,5×38,1 mm (2,5×1,5")
    1000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 824,00 kr
    824,00 kr
    1 * 1 ROL
       824,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-238C1-1WH
  • GAINJTTA-11C1-2WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    White
    67,1×7 mm (2,64×0,28")
    2000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 601,00 kr
    601,00 kr
    1 * 1 ROL
       601,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-11C1-2WH
  • GAINJTTA-181C1-1WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    White
    70×38,1 mm (2,75×1,5")
    1000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 903,00 kr
    903,00 kr
    1 * 1 ROL
       903,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-181C1-1WH
  • GAINJTTA-181C3-2WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    White
    70×38,1 mm (2,75×1,5")
    2000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 1.810,00 kr
    1.810,00 kr
    1 * 1 ROL
       1.810,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-181C3-2WH
  • GAINJTTA-36C1-1WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    White
    76,2×25,4 mm (3×1")
    1000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 682,00 kr
    682,00 kr
    1 * 1 ROL
       682,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-36C1-1WH
  • GAINJTTA-36C3-1WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    White
    76,2×25,4 mm (3×1")
    1000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 682,00 kr
    682,00 kr
    1 * 1 ROL
       682,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-36C3-1WH
  • GAINJTTA-56C1-1WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    White
    76,2×50,8 mm (3×2")
    1000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 1.290,00 kr
    1.290,00 kr
    1 * 1 ROL
       1.290,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-56C1-1WH
  • GAINJTTA-56C3-1WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    White
    76,2×50,8 mm (3×2")
    1000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 1.290,00 kr
    1.290,00 kr
    1 * 1 ROL
       1.290,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-56C3-1WH
  • GAINJTTA99C10.25WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    White
    76,2×76,2 mm (3×3")
    250 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 472,00 kr
    472,00 kr
    1 * 1 ROL
       472,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA99C10.25WH
  • GAINJTTA-101C1-1WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    White
    76×38,1 mm (3×1,5")
    1000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 984,00 kr
    984,00 kr
    1 * 1 ROL
       984,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-101C1-1WH
  • GAINJTTA122C10.3WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    White
    101,6×101,6 mm (4×4")
    300 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 987,00 kr
    987,00 kr
    1 * 1 ROL
       987,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA122C10.3WH
  • GAINJTTA-127C1-1WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    White
    101,6×25,4 mm (4×1")
    1000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 896,00 kr
    896,00 kr
    1 * 1 ROL
       896,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-127C1-1WH
  • GAINJTTA-127C3-1WH
    Rectangle permanent cryogenic barcode labels, core size 3"
    Rectangle permanent cryogenic barcode labels, core size 3"
    White
    101,6×25,4 mm (4×1")
    1000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 896,00 kr
    896,00 kr
    1 * 1 ROL
       896,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA-127C3-1WH
  • GAINJTTA62C1-0.5WH
    Rectangle permanent cryogenic barcode labels, core size 1"
    Rectangle permanent cryogenic barcode labels, core size 1"
    White
    101,6×50,8 mm (4×2")
    500 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 ROL
    1 * 1 ROL - 847,00 kr
    847,00 kr
    1 * 1 ROL
       847,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    GAINJTTA62C1-0.5WH