Cryogenic Tamper-Evident Tapes
: GA
This tamper-evident tape is designed for use with cryogenic containers such as cryo vials, cryo/freezer boxes, cryo cassettes, and tubes.
- Temperature range: −196 to +110 °C (−320,8 to 230 °F)
- Able to withstand immersion in liquid nitrogen and deep freeze storage conditions
- Continuous tape format allows use of as much or as little as necessary
- Available in assorted colours for easy identification and colour coding
The continuous tape will tear-up upon any attempt to remove it from most surfaces, indicating whether the contents have been compromised. Durable and waterproof, the tamper-evident tape is available in a range of colours for easy identification. Writable using permanent cryogenic markers, the cryo tape is suitable for long-term storage in liquid nitrogen (−196 °C), laboratory freezers (−80 °C), and for transportation on dry ice.
Frequently Bought Together