Air compressor, corona
Corona air compressor
Catalog # 564-0299
Supplier: PEAK SCIENTIFIC
Jump to:
Product Details & Documents
The corona air compressor has been engineered to stack neatly with the corona 1010 nitrogen generator providing compressed air for laboratories without an external source of gas. The corona series has been developed to meet the requirements of the thermo scientific corona veo Charged aerosol detector.
- Stackable with corona 1010 nitrogen generator
- Provides air for corona system for laboratories without an external supply
- Gas is supplied on demand so generator works to your schedule
- 1 year comprehensive on-site warranty
Specifications
- Description:Corona air compressor