Order Entry
Denmark
ContactUsLinkComponent
n-Pentane 99% (by GC), AR®, Macron Fine Chemicals™
Flammable
Irritant
Health
Environment
n-Pentane 99% (by GC), AR®, Macron Fine Chemicals™
Certificates
Flammable
Irritant
Health
Environment
n-Pentane 99% (by GC), AR®, Macron Fine Chemicals™
Certificates

Specification Test Results

  • Assay (GC):
    ≥99,0%
  • Water (KF):
    ≤0,02%
  • Titrable Acid (meq/g):
    ≤0,0005
  • Residue after Evaporation:
    ≤0,001%
  • Sulfur Compounds:
    ≤0,001%

Specification Test Results

Related Information

Frequently Bought Together