Order Entry
Denmark
ContactUsLinkComponent
Lithium hydroxide, anhydrous ≥99%, AnalaR NORMAPUR®
Corrosive
Toxic
Lithium hydroxide, anhydrous ≥99%, AnalaR NORMAPUR®
Supplier:  VWR Chemicals
Certificates
undefined
Corrosive
Toxic
Lithium hydroxide, anhydrous ≥99%, AnalaR NORMAPUR®
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    min. 99%
  • Heavy metals (as Pb):
    max. 20 ppm
  • Lithium carbonate:
    max. 1.0%
  • Cl (Chloride):
    max. 0.04%
  • SO4 (Sulphate):
    max. 0.04%
  • Ca (Calcium):
    max. 0.02%
  • Fe (Iron):
    max. 20 ppm
  • K (Potassium):
    max. 0.02%
  • Na (Sodium):
    max. 0.05%

Specification Test Results

Related Information

Frequently Bought Together