Celite® 512, medium filter aid, GPR RECTAPUR®
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:91053-39-3
:61790-53-2
Formula:SiO₂
:60.08 g/mol
Specification Test Results
- pH (25 °C; 10%):8.5 - 11.0
- Wet density:≤ 370 g/l
- Loss on ignition (1000 °C):≤ 0.4%
- Particle size (< 75µm):≥ 90%
Specification Test Results
Related Information
Certificates
Search by Lot Numberspec_VWRC22555.290 spec_VWRC22555.360
Product specification
VWR® for Drying and Filtration
Product Literature
Frequently Bought Together