Pressure booster
Unipump® I DC, 219×390×214 mm, −120...300 °C (temperature range)
Catalog # HUBE1085.0001.00
Supplier: HUBER
Jump to:
Product Details & Documents
The stainless steel Unipump® is designed to compensate for pressure loss in external systems.
- Operation either manual (on/off) or external via 3-line-connector
The Unipump® is connected in series with the pump of the Unistat, Unichiller or Compatible Control circulator and can be controlled via the voltfree contact of the Com.Gate (Option).
Pump connection: M24×1,5
Max. pump capacities: 55 L/min, 1000 mbar
Ordering information: Accessories included. There are also some optional accessories.
Specifications
- Description:Unipump® I DC, 219×390×214 mm, −120...300 °C (temperature range)
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



