Order Entry
Denmark
ContactUsLinkComponent
Bottles, amber, glass, with thread, Assistent®
Bottles, amber, glass, with thread, Assistent®
Catalog # HECH40190030
undefined
Bottles, amber, glass, with thread, Assistent®
Catalog # HECH40190030
Specifications
  • Capacity:
    1000 ml
  • Colour:
    Amber
  • Description:
    Glass bottle with thread
  • Seq:
    02
  • Pk:
    1
Specifications
About this item
Amber glass finish is designed to protect light-sensitive samples in transport and storage.
  • Space saving square design
  • With screw cap
Bottles are supplied with a screw cap made of polypropylene, which is resistant to steam sterilsation up to 121 °C (249,8 °F).