Hermetic Tamper Evident Buckets, Round
Robust PP bucket with ergonomic handle. It features pressure lid with opening-off seal.
- Flat bottom ensures total emptying and easy cleaning
- Especially suitable in cosmetic, pharma, paint or dye industries
- Food contact material approved
Hot filling possible up to 95 °C.
Frequently Bought Together