Order Entry
Denmark
Orders LinkContactUsLinkComponent
ICP-MS interference check mixes
ICP-MS interference check mixes
ICP-MS interference check mixes
Certificates
ICP-MS interference check mixes
ICP-MS interference check mixes
2 Options Available Below
Product Details & Documents
It offers a comprehensive range of single and multi-element standards, tuning solutions, ionisation buffers and environmental standards for atomic spectroscopy.
  • It is works in AAS, ICP-OES or ICP-MS and MP-AES results
  • Accurate, reliable calibrations
Standards are prepared to exacting specifications that ensure consistency. Every solution is manufactured using high purity starting materials, which are regularly tested in accordance with NIST guidelines to provide traceability.
Product Family Options
Product Information
  • Matrix
    Pk
    Availability
    Price
  • HEWL5188-6526
    6020 Interference check solution A. For use with ICP-MS systems. Contains: 20000 ppm Cl, 3000 ppm Ca, 2500 ppm Fe and Na, 2000 ppm C, 1000 ppm Al, Mg, P, K, S and 20 ppm Mo and Ti. 100 ml
    6020 Interference check solution A. For use with ICP-MS systems. Contains: 20000 ppm Cl, 3000 ppm Ca, 2500 ppm Fe and Na, 2000 ppm C, 1000 ppm Al, Mg, P, K, S and 20 ppm Mo and Ti. 100 ml
    5% HNO₃
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 5.670,00 kr
    5.670,00 kr
    1 * 100 mL
       5.670,00 kr
  • HEWL5188-6527
    6020 Interference check solution B. For use with ICP-MS systems. Contains: 20 ppm of Cr, Co, Cu, Mn, Ni and V, 10 ppm of As, Cd, Se, and Zn, 5 ppm of Ag. Bottle of 100 ml
    6020 Interference check solution B. For use with ICP-MS systems. Contains: 20 ppm of Cr, Co, Cu, Mn, Ni and V, 10 ppm of As, Cd, Se, and Zn, 5 ppm of Ag. Bottle of 100 ml
    5% HNO₃
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 2.320,00 kr
    2.320,00 kr
    1 * 1 Items
       2.320,00 kr