Order Entry
Denmark
ContactUsLinkComponent
Proteinase K
Proteinase K
Catalog # AGLS300140
Supplier:  AGILENT
CAS Number:  
Proteinase K
Catalog # AGLS300140
Supplier:  AGILENT
CAS Number:  
Specifications
  • Pk:
    1
  • Enzyme name:
    Proteinase K
Specifications
About this item
Proteinase K is a broad spectrum protease from Tritirachium album that is ideal for preparing chromosomal DNA.
  • For general digestion of protein in biological samples
  • Lyophilised powder with specific activity >30 units/mg at 37 °C
  • Size: 100 mg
It cleaves peptide bonds in which carbonyl groups are surrounded by aromatic, hydrophobic, or aliphatic amino acids.
Used for the following applications: pulsed field electrophoresis, chromosomal DNA for protein fingerprinting, and nuclease removal from DNA and RNA preparations.
Caution: For research use only. Not for use in diagnostic procedures.