Order Entry
Denmark
Orders LinkContactUsLinkComponent
TE Buffers 100X
Supplier:   G-Biosciences
TE Buffers 100X
Supplier:  G-Biosciences
Certificates
3 Options Available Below
Product Details & Documents
TE Buffer (Tris-EDTA Buffer) is a common molecular biology buffer that is used to protect DNA and RNA from degradation. The Tris buffering agent and EDTA metal chelating properties help protect DNA and RNA.
Product Family Options
Product Information
  • pH value
    Pack type
    Pk
    Availability
    Price
  • R003
    TE buffer, 100X
    TE buffer, 100X
    8,0
    Plastic bottle
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 344,00 kr
    344,00 kr
    1 * 100 mL
       344,00 kr
  • 786-752
    TE buffer, 100X
    TE buffer, 100X
    8,0
    Plastic bottle
    250 mL
    Log in for availability
    Log in for availability
    1 * 250 mL
    1 * 250 mL - 653,00 kr
    653,00 kr
    1 * 250 mL
       653,00 kr
  • 786-813
    TE buffer, 100X
    TE buffer, 100X
    8,0
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 1.160,00 kr
    1.160,00 kr
    1 * 500 mL
       1.160,00 kr