Order Entry
Denmark
ContactUsLinkComponent
Human recombinant Matrix Metalloproteinase-7
Human recombinant Matrix Metalloproteinase-7
Supplier:  Apollo Scientific
Certificates
Human recombinant Matrix Metalloproteinase-7
Supplier:  Apollo Scientific

Specifications

  • Enzyme type:
    Recombinant
  • Species:
    Human
  • Storage conditions:
    Ambient
  • MDL No.:
    MFCD01633460
  • Enzyme name:
    Matrix Metalloproteinase-7
  • Enzyme synonyms:
    MMP-7

Specifications

Frequently Bought Together