Order Entry
Denmark
ContactUsLinkComponent
Recombinant isocitrate dehydrogenase
Recombinant isocitrate dehydrogenase
Supplier:  Apollo Scientific
Certificates
Recombinant isocitrate dehydrogenase
Supplier:  Apollo Scientific

Specifications

  • Enzyme type:
    Recombinant
  • Storage conditions:
    Refrigerator
  • MDL No.:
    MFCD00131420
  • Enzyme name:
    Isocitrate Dehydrogenase
  • Enzyme synonyms:
    iCDH

Specifications

Frequently Bought Together