Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Denmark
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Enzymes
Recombinant disulfide bond isomerase
Recombinant disulfide bond isomerase
Recombinant disulfide bond isomerase
APOSBITP1319-10UG
:
Apollo Scientific
:
Recombinant disulfide bond isomerase
APOSBITP1319-10UG
:
Apollo Scientific
:
Pk:
10 µG
Enzyme type:
Recombinant
Storage conditions:
Freezer
Enzyme name:
Disulfide Bond Isomerase
Enzyme synonyms:
DsbC