Order Entry
Denmark
ContactUsLinkComponent
Recombinant beta lactamase
Recombinant beta lactamase
Supplier:  Apollo Scientific
Certificates
Recombinant beta lactamase
Supplier:  Apollo Scientific

Specifications

  • Enzyme type:
    Recombinant
  • Storage conditions:
    Ambient
  • Enzyme name:
    Beta Lactamase
  • Enzyme synonyms:
    b-Lactamase

Specifications

Frequently Bought Together