Order Entry
Denmark
Orders LinkContactUsLinkComponent
Recombinant collagenase H

Recombinant collagenase H

Catalog #
Recombinant collagenase H
Catalog #
Supplier:  
CAS Number:  
Recombinant collagenase H
CAS Number:  
Product Details & Documents

From Clostridium histolyticum and produced in E. Coli and B. Megaterium. Endotoxin negative. No detectable Caseinase activity. Sterile lyophilized powder.