Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Denmark
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Antibodies
Anti-HA Tag Mouse Antibody
Anti-HA Tag Mouse Antibody
Anti-HA Tag Mouse Antibody
APOSBITP2143-0.5MG
:
Apollo Scientific
Anti-HA Tag Mouse Antibody
APOSBITP2143-0.5MG
:
Apollo Scientific
Antigen symbol:
HA Tag
Host:
Mouse
Pk:
0,5 mg