0,00 kr
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Denmark
ContactUsLinkComponent
Antibodies
Anti-NG Mouse Antibody
Anti-NG Mouse Antibody
APOSBITP2049-1MG
:
Apollo Scientific
Anti-NG Mouse Antibody
APOSBITP2049-1MG
:
Apollo Scientific
Antigen name:
Neisseria Gonorrhea
Antigen symbol:
NG
Host:
Mouse
Isotype:
Reactivity:
Environmentally Preferable:
Pk:
1 mg