Order Entry
Denmark
ContactUsLinkComponent
Anti-B220 Rat Antibody
Anti-B220 Rat Antibody
Catalog # APOSBITP2027-1MG
Supplier:  Apollo Scientific
Anti-B220 Rat Antibody
Catalog # APOSBITP2027-1MG
Supplier:  Apollo Scientific

Specifications

  • Antigen symbol:
    B220
  • Host:
    Rat
  • Reactivity:
    Mouse
  • Pk:
    1 mg

Specifications

About this item