Order Entry
Denmark
ContactUsLinkComponent
Recombinant Dengue Virus NS1 c-end Type 2
Recombinant Dengue Virus NS1 c-end Type 2
Catalog # APOSBITP1960-1MG
Supplier:  Apollo Scientific
CAS Number:  
Recombinant Dengue Virus NS1 c-end Type 2
Catalog # APOSBITP1960-1MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Protein/peptide type:
    Recombinant
  • Protein synonyms:
    Dengue NS1c
  • Protein/peptide name:
    Dengue Virus NS1 c-end Type 2

Specifications

About this item