Recombinant Epstein-Barr Virus (HHV-4) p23
Catalog # APOSBITP1918-1MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pk:1 mg
- Protein/peptide type:Recombinant
- Protein synonyms:EBV p23
- Protein/peptide name:Epstein-Barr Virus (HHV-4) p23