Order Entry
Denmark
ContactUsLinkComponent
 
Recombinant Toxoplasma Gondii P30 (SAG 1)
Catalog # APOSBITP1852-1MG
Supplier:  Apollo Scientific
CAS Number:  
Recombinant Toxoplasma Gondii P30 (SAG 1)
Catalog # APOSBITP1852-1MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Protein/peptide type:
    Recombinant
  • Protein synonyms:
    Toxoplasma P30
  • Protein/peptide name:
    Toxoplasma Gondii P30 (SAG 1)

Specifications

About this item