Order Entry
Denmark
ContactUsLinkComponent
Recombinant Toxoplasma Gondii P29 (GRA 7)
Recombinant Toxoplasma Gondii P29 (GRA 7)
Catalog # APOSBITP1851-1MG
Supplier:  Apollo Scientific
CAS Number:  
Recombinant Toxoplasma Gondii P29 (GRA 7)
Catalog # APOSBITP1851-1MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Protein/peptide type:
    Recombinant
  • Protein synonyms:
    Toxoplasma P29
  • Protein/peptide name:
    Toxoplasma Gondii P29 (GRA 7)

Specifications

About this item