0,00 kr
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Denmark
ContactUsLinkComponent
Proteins and Peptides
Recombinant Hepatitis C Virus NS5 Genotype-1b
Recombinant Hepatitis C Virus NS5 Genotype-1b
APOSBITP1797-1MG
:
Apollo Scientific
:
Recombinant Hepatitis C Virus NS5 Genotype-1b
APOSBITP1797-1MG
:
Apollo Scientific
:
Pk:
1 mg
Protein/peptide type:
Recombinant
Protein synonyms:
HCV NS5 Genotype-1b
Protein/peptide name:
Hepatitis C Virus NS5 Genotype-1b