Recombinant Hepatitis C Virus NS3, Biotin
Catalog # APOSBITP1781-1MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pk:1 mg
- Conjugation:Biotin
- Protein/peptide type:Recombinant
- Protein synonyms:HCV NS3
- Protein/peptide name:Hepatitis C Virus NS3