Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Denmark
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Proteins and Peptides
Recombinant Hepatitis C Virus Combined
Recombinant Hepatitis C Virus Combined
Recombinant Hepatitis C Virus Combined
Catalog # APOSBITP1758-1MG
Supplier:
Apollo Scientific
CAS Number:
Expand image
Recombinant Hepatitis C Virus Combined
Catalog # APOSBITP1758-1MG
Supplier:
Apollo Scientific
CAS Number:
Specifications
Pk:
1 mg
Protein/peptide type:
Recombinant
Protein synonyms:
HCV Combined
Protein/peptide name:
Hepatitis C Virus Combined
Specifications
About this item
Product Details
Product Details