Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Denmark
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Proteins and Peptides
Recombinant Herpes Simplex Virus-2 gG
Recombinant Herpes Simplex Virus-2 gG
Recombinant Herpes Simplex Virus-2 gG
APOSBITP1739-1MG
:
Apollo Scientific
:
Recombinant Herpes Simplex Virus-2 gG
APOSBITP1739-1MG
:
Apollo Scientific
:
Pk:
1 mg
Protein/peptide type:
Recombinant
Protein synonyms:
rHSV-2 gG
Protein/peptide name:
Herpes Simplex Virus-2 gG