Order Entry
Denmark
ContactUsLinkComponent
Recombinant SARS Associated Coronavirus Nucleocaspid (1-49,192-220)
Recombinant SARS Associated Coronavirus Nucleocaspid (1-49,192-220)
Catalog # APOSBITP1730-1MG
Supplier:  Apollo Scientific
CAS Number:  
Recombinant SARS Associated Coronavirus Nucleocaspid (1-49,192-220)
Catalog # APOSBITP1730-1MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Protein/peptide type:
    Recombinant
  • Protein synonyms:
    SARS Nucleocaspid
  • Protein/peptide name:
    SARS Associated Coronavirus Nucleocaspid (1-49,192-220)
  • Sequence:
    1-49; 192-220

Specifications

About this item