Order Entry
Denmark
ContactUsLinkComponent
Human recombinant superoxide dismutase
Human recombinant superoxide dismutase
Catalog # APOSBITP1650-1MG
Supplier:  Apollo Scientific
CAS Number:  
Human recombinant superoxide dismutase
Catalog # APOSBITP1650-1MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Seq:
    23
  • Protein synonyms:
    SOD
  • Protein/peptide name:
    Superoxide Dismutase

Specifications

About this item