Human Recombinant Creatine Kinase MB Isoenzyme Type-2
Catalog # APOSBITP1435-1MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pk:1 mg
- Protein/peptide type:Recombinant
- Species:Human
- Protein synonyms:CKMB Type-2
- Protein/peptide name:Creatine Kinase MB Isoenzyme Type-2