Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Denmark
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Hjem
Proteins and Peptides
Mouse recombinant MCP-1
Mouse recombinant MCP-1
Mouse recombinant MCP-1
APOSBITC2025-1MG
:
Apollo Scientific
:
Mouse recombinant MCP-1
APOSBITC2025-1MG
:
Apollo Scientific
:
Pk:
1 mg
Protein/peptide type:
Recombinant
Species:
Mouse
Protein synonyms:
Monocyte Chemotactic Protein-1, CCL2
Protein/peptide name:
MCP-1