Waste caps, b.safe
About this item
Safe collection of hazardous waste is an important part of the laboratory work routine. b.safe waste caps connect waste lines from your HPLC/UHPLC leak-tight with your waste container. The waste caps are available in various sizes to fit your specific waste container safely.
- Available in many thread sizes to fit a wide range of waste containers
- Excellent chemical resistance
- Accessories like fittings and hose connectors guarantee you waste lines are connected leak tight
- Special couplings for corrugated flexible tubes
- Exhaust filters for different sizes of waste containers that adsorb hazardous vapours
Suitable fittings, hose connectors and special couplings for corrugated hoses, make the b.safe waste system very flexible. Materials like PTFE, PPS, PFA and PP guarantee excellent chemical resistant. Now all you need is a b.safe exhaust filter to complete your safe waste collection system.
Safe collection of harmful solvent waste as well as perfect protection from hazardous vapours and liquids, also absolute control until non-hazardous collection of waste. This is what b.safe stands for.
After extraction and analysis have been successfully completed, lab experts cannot relax because at the end of every HPLC process, there is harmful solvent waste that increases the risk of fire and explosion and the health of employees. The waste has, therefore, to be collected in special vessels until their disposal without vapours and liquids being able to escape. A clear case for b.safe waste caps and b.safe exhaust filters.
b.safe waste caps are fixed tightly on the threads of the waste containers. Fittings, hose connectors and the practical distributor body, tubing and capillaries can be fixed without the unit being dismantled. As a result, eluents come into the waste containers safely and not outside of them. A special activated carbon in the b.safe exhaust filters absorbs all solvent vapours in order to have only harmless air coming into the environment. Practical indicators guarantee full control on permitted service life.
37 Options Available Below
Product Details & Documents
Safe collection of hazardous waste is an important part of the laboratory work routine. b.safe waste caps connect waste lines from your HPLC/UHPLC leak-tight with your waste container. The waste caps are available in various sizes to fit your specific waste container safely.
- Available in many thread sizes to fit a wide range of waste containers
- Excellent chemical resistance
- Accessories like fittings and hose connectors guarantee you waste lines are connected leak tight
- Special couplings for corrugated flexible tubes
- Exhaust filters for different sizes of waste containers that adsorb hazardous vapours
Suitable fittings, hose connectors and special couplings for corrugated hoses, make the b.safe waste system very flexible. Materials like PTFE, PPS, PFA and PP guarantee excellent chemical resistant. Now all you need is a b.safe exhaust filter to complete your safe waste collection system.
Safe collection of harmful solvent waste as well as perfect protection from hazardous vapours and liquids, also absolute control until non-hazardous collection of waste. This is what b.safe stands for.
After extraction and analysis have been successfully completed, lab experts cannot relax because at the end of every HPLC process, there is harmful solvent waste that increases the risk of fire and explosion and the health of employees. The waste has, therefore, to be collected in special vessels until their disposal without vapours and liquids being able to escape. A clear case for b.safe waste caps and b.safe exhaust filters.
b.safe waste caps are fixed tightly on the threads of the waste containers. Fittings, hose connectors and the practical distributor body, tubing and capillaries can be fixed without the unit being dismantled. As a result, eluents come into the waste containers safely and not outside of them. A special activated carbon in the b.safe exhaust filters absorbs all solvent vapours in order to have only harmless air coming into the environment. Practical indicators guarantee full control on permitted service life.
Documents
Manual b.safe Waste Caps
Manual
TDS b.safe Waste Cap
Datasheet
Product Family Options
Product Information
- DescriptionThreadTilgængelighedPrice
- Waste caps, 3× capillary connector, including 3× fittings for tubing with ext. Ø 1,6 mm, 3× fittings for tubing with ext. Ø 2,2 mm, 3× fittings for tubing with ext. Ø 3,2 mm, 2 blind fittingsGL45Log in for availability1 * 1 Items897,00 kr
- Waste caps, 2× capillary connector, including 2× fittings for tubing with ext. Ø 1,6 mm, 2× fittings for tubing with ext. Ø 2,2 mm, 2× fittings for tubing with ext. Ø 3,2 mm, 1 blind fittings, 1 hose connector for tubing with int. Ø 5,5 - 8,0 mmGL45Log in for availability1 * 1 Items790,00 kr
- Waste caps, 4× capillary connector, including 4× fittings for tubing with ext. Ø 1,6 mm, 4× fittings for tubing with ext. Ø 2,2 mm, 4× fittings for tubing with ext. Ø 3,2 mm, 3 blind fittings, 1 hose connector for tubing with int. Ø 5,5 - 8,0 mmGL45Log in for availability1 * 1 Items1.190,00 kr
- Waste cap with funnel, 1× capillary connector, including 1× fitting for tubing with ext. Ø 1,6 mm, 1× fitting for tubing with ext. Ø 2,2 mm, 1× fitting for tubing with ext. Ø 3,2 mm, 1 blind fitting, 1 hose connector for tubing with int. Ø 5,5 - 8,0 mm, 1 funnel with automatic stopcockGL45Log in for availability1 * 1 Items4.270,00 kr
- Waste cap with funnel, 1× capillary connector, including 1× fitting for tubing with ext. Ø 1,6 mm, 1× fitting for tubing with ext. Ø 2,2 mm, 1× fitting for tubing with ext. Ø 3,2 mm, 1 blind fitting, 1 hose connector for tubing with int. Ø 5,5 - 8,0 mm, 1x funnel with stopcock with gripGL45Log in for availability1 * 1 Items4.650,00 kr
- Waste caps, 3× capillary connector, including 3× fittings for tubing with ext. Ø 1,6 mm, 3× fittings for tubing with ext. Ø 2,2 mm, 3× fittings for tubing with ext. Ø 3,2 mm, 2 blind fittingsS40Log in for availability1 * 1 Items998,00 kr
- Waste caps, 2× capillary connector, including 2× fittings for tubing with ext. Ø 1,6 mm, 2× fittings for tubing with ext. Ø 2,2 mm, 2× fittings for tubing with ext. Ø 3,2 mm, 1 blind fitting, 1 hose connector for tubing with int. Ø 5,5 - 8,0 mmS40Log in for availability1 * 1 Items919,00 kr
- Waste caps, 3× capillary connector, including 3× fittings for tubing with ext. Ø 1,6 mm, 3× fittings for tubing with ext. Ø 2,2 mm, 3× fittings for tubing with ext. Ø 3,2 mm, 2 blind fittingsS50Log in for availability1 * 1 Items1.220,00 kr
- Waste caps, 2× capillary connector, including 2× fittings for tubing with ext. Ø 1,6 mm, 2× fittings for tubing with ext. Ø 2,2 mm, 2× fittings for tubing with ext. Ø 3,2 mm, 1 blind fitting, 1 hose connector for tubing with int. Ø 5,5 - 8,0 mmS50Log in for availability1 * 1 Items1.350,00 kr
- Waste caps, 4× capillary connector, including 4× fittings for tubing with ext. Ø 1,6 mm, 4× fittings for tubing with ext. Ø 2,2 mm, 4× fittings for tubing with ext. Ø 3,2 mm, 8 blind fittings, 4 hose connectors for tubing with int. Ø 5,5 - 8,0 mmS50Log in for availability1 * 1 Items1.640,00 kr
- Waste caps, 3× capillary connector, including 3× fittings for tubing with ext. Ø 1,6 mm, 3× fittings for tubing with ext. Ø 2,2 mm, 3× fittings for tubing with ext. Ø 3,2 mm, 2 blind fittingsS51Log in for availability1 * 1 Items1.280,00 kr
- Waste caps, 2× capillary connector, including 2× fittings for tubing with ext. Ø 1,6 mm, 2× fittings for tubing with ext. Ø 2,2 mm, 2× fittings for tubing with ext. Ø 3,2 mm, 1 blind fitting, 1 hose connector for tubing with int. Ø 5,5 - 8,0 mmS51Log in for availability1 * 1 Items1.420,00 kr
- Waste cap with funnel, 1× capillary connector, including 1× fitting for tubing with ext. Ø 1,6 mm, 1× fitting for tubing with ext. Ø 2,2 mm, 1× fitting for tubing with ext. Ø 3,2 mm, 1 hose connector for tubing with int. Ø 5,5 - 8,0 mm, 1 blind fitting, 1x funnel with stopcockS51Log in for availability1 * 1 Items4.270,00 kr
- Waste caps, 3× capillary connector, including 3× fittings for tubing with ext. Ø 1,6 mm, 3× fittings for tubing with ext. Ø 2,2 mm, 3× fittings for tubing with ext. Ø 3,2 mm, 2 blind fittingsS55Log in for availability1 * 1 Items1.210,00 kr
- Waste caps, 2× capillary connector, including 2× fittings for tubing with ext. Ø 1,6 mm, 2× fittings for tubing with ext. Ø 2,2 mm, 2× fittings for tubing with ext. Ø 3,2 mm, 1 blind fitting, 1 hose connector for tubing with int. Ø 5,5 - 8,0 mmS55Log in for availability1 * 1 Items1.210,00 kr
- Waste caps, 4× capillary connector, including 4× fittings for tubing with ext. Ø 1,6 mm, 4× fittings for tubing with ext. Ø 2,2 mm, 4× fittings for tubing with ext. Ø 3,2 mm, 8 blind fittings, 4 hose connectors for tubing with int. Ø 5,5 - 8,0 mmS55Log in for availability1 * 1 Items1.360,00 kr
- Waste cap with funnel, 2× capillary connector, including 2× fittings for tubing with ext. Ø 1,6 mm, 2× fittings for tubing with ext. Ø 2,2 mm, 2× fittings for tubing with ext. Ø 3,2 mm, 1 blind fitting, 1 hose connector for tubing with int. Ø 5,5 - 8,0 mm, 1 funnel with automatic stopcockS55Log in for availability1 * 1 Items3.880,00 kr
- Waste cap with funnel, 2× capillary connector, including 2× fittings for tubing with ext. Ø 1,6 mm, 2× fittings for tubing with ext. Ø 2,2 mm, 2× fittings for tubing with ext. Ø 3,2 mm, 1 blind fitting, 1 hose connector for tubing with int. Ø 5,5 - 8,0 mm, 1x funnel with stopcock with gripS55Log in for availability1 * 1 Items4.630,00 kr
- Waste caps, 3× capillary connector, including 3× fittings for tubing with ext. Ø 1,6 mm, 3× fittings for tubing with ext. Ø 2,2 mm, 3× fittings for tubing with ext. Ø 3,2 mm, 2 blind fittingsS60Log in for availability1 * 1 Items1.320,00 kr
- Waste caps, 2× capillary connector, including 2× fittings for tubing with ext. Ø 1,6 mm, 2× fittings for tubing with ext. Ø 2,2 mm, 2× fittings for tubing with ext. Ø 3,2 mm, 1 blind fitting, 1 hose connector for tubing with int. Ø 5,5 - 8,0 mmS60Log in for availability1 * 1 Items1.270,00 kr
- Waste caps, 4× capillary connector, including 4× fittings for tubing with ext. Ø 1,6 mm, 4× fittings for tubing with ext. Ø 2,2 mm, 4× fittings for tubing with ext. Ø 3,2 mm, 8 blind fittings, 4 hose connectors for tubing with int. Ø 5,5 - 8,0 mmS60Log in for availability1 * 1 Items1.380,00 kr
- Waste cap with funnel, 2× capillary connector, including 2× fittings for tubing with ext. Ø 1,6 mm, 2× fittings for tubing with ext. Ø 2,2 mm, 2× fittings for tubing with ext. Ø 3,2 mm, 1 blind fittings, 1 hose connector for tubing with int. Ø 5,5 - 8,0 mm, 1 funnel with automatic stopcockS60Log in for availability1 * 1 Items3.890,00 kr
- Waste cap with funnel, 2× capillary connector, including 2× fittings for tubing with ext. Ø 1,6 mm, 2× fittings for tubing with ext. Ø 2,2 mm, 2× fittings for tubing with ext. Ø 3,2 mm, 1 blind fitting, 1 hose connector for tubing with int. Ø 5,5 - 8,0 mm, 1x funnel with stopcock with gripS60Log in for availability1 * 1 Items4.670,00 kr
- Waste caps, 5× capillary connector, including 5× fittings for tubing with ext. Ø 1,6 mm, 5× fittings for tubing with ext. Ø 2,2 mm, 5× fittings for tubing with ext. Ø 3,2 mm, 4 blind fittingsS65Log in for availability1 * 1 Items1.550,00 kr
- Waste caps, 4× capillary connector, including 4× fittings for tubing with ext. Ø 1,6 mm, 4× fittings for tubing with ext. Ø 2,2 mm, 4× fittings for tubing with ext. Ø 3,2 mm, 3 blind fittings, 1 hose connectors for tubing with int. Ø 5,5 - 8,0 mmS65Log in for availability1 * 1 Items1.600,00 kr
- Waste caps, 4× capillary connector, including 4× fittings for tubing with ext. Ø 1,6 mm, 4× fittings for tubing with ext. Ø 2,2 mm, 4× fittings for tubing with ext. Ø 3,2 mm, 3 blind fittingsS70Log in for availability1 * 1 Items1.800,00 kr
- Waste caps, 4× capillary connector, including 4× fittings for tubing with ext. Ø 1,6 mm, 4× fittings for tubing with ext. Ø 2,2 mm, 4× fittings for tubing with ext. Ø 3,2 mm, 3 blind fittings, 1 hose connectors for tubing with int. Ø 5,5 - 8,0 mmS70Log in for availability1 * 1 Items1.800,00 kr
- Waste cap with funnel, 3× capillary connector, including 3× fittings for tubing with ext. Ø 1,6 mm, 3× fittings for tubing with ext. Ø 2,2 mm, 3× fittings for tubing with ext. Ø 3,2 mm, 2 blind fittings, 1 hose connector for tubing with int. Ø 5,5 - 8,0 mm, 1x funnel with stopcockS70Log in for availability1 * 1 Items3.050,00 kr
- Waste caps, 4× capillary connector, including 4× fittings for tubing with ext. Ø 1,6 mm, 4× fittings for tubing with ext. Ø 2,2 mm, 4× fittings for tubing with ext. Ø 3,2 mm, 3 blind fittingsS90Log in for availability1 * 1 Items2.010,00 kr
- Waste caps, 4× capillary connector, including 4× fittings for tubing with ext. Ø 1,6 mm, 4× fittings for tubing with ext. Ø 2,2 mm, 4× fittings for tubing with ext. Ø 3,2 mm, 3 blind fittings, 1 hose connectors for tubing with int. Ø 5,5 - 8,0 mmS90Log in for availability1 * 1 Items2.170,00 kr
- Waste caps, 4× capillary connector, including 4× fittings for tubing with ext. Ø 1,6 mm, 4× fittings for tubing with ext. Ø 2,2 mm, 4× fittings for tubing with ext. Ø 3,2 mm, 3 blind fittings, 1 hose connectors for tubing with int. Ø 5,5 - 8,0 mmB53Log in for availability1 * 1 Items2.340,00 kr
- Waste cap with funnel, 2× capillary connector, including 2× fittings for tubing with ext. Ø 1,6 mm, 2× fittings for tubing with ext. Ø 2,2 mm, 2× fittings for tubing with ext. Ø 3,2 mm, 1 blind fitting, 1 hose connector for tubing with int. Ø 5,5 - 8,0 mm, 1 funnel with automatic stopcockB53Log in for availability1 * 1 Items5.380,00 kr
- Waste cap with funnel, 2× capillary connector, including 2× fittings for tubing with ext. Ø 1,6 mm, 2× fittings for tubing with ext. Ø 2,2 mm, 2× fittings for tubing with ext. Ø 3,2 mm, 1 blind fitting, 1 hose connector for tubing with int. Ø 5,5 - 8,0 mm, 1x funnel with stopcock with gripB53Log in for availability1 * 1 Items3.700,00 kr
- Waste caps, 3× capillary connector, including 3× fittings for tubing with ext. Ø 1,6 mm, 3× fittings for tubing with ext. Ø 2,2 mm, 3× fittings for tubing with ext. Ø 3,2 mm, 2 blind fittingsB63Log in for availability1 * 1 Items2.510,00 kr
- Waste caps, 2× capillary connector, including 2× fittings for tubing with ext. Ø 1,6 mm, 2× fittings for tubing with ext. Ø 2,2 mm, 2× fittings for tubing with ext. Ø 3,2 mm, 1 blind fitting, 1 hose connector for tubing with int. Ø 5,5 - 8,0 mmB63Log in for availability1 * 1 Items2.170,00 kr
- Waste caps, 3× capillary connector, including 3× fittings for tubing with ext. Ø 1,6 mm, 3× fittings for tubing with ext. Ø 2,2 mm, 3× fittings for tubing with ext. Ø 3,2 mm, 2 blind fittings, 1 hose connector for tubing with int. Ø 5,5 - 8,0 mmB63Log in for availability1 * 1 Items2.110,00 kr
- Waste caps, 4× capillary connector, including 4× fittings for tubing with ext. Ø 1,6 mm, 4× fittings for tubing with ext. Ø 2,2 mm, 4× fittings for tubing with ext. Ø 3,2 mm, 3 blind fittings, 1 hose connectors for tubing with int. Ø 5,5 - 8,0 mmB63Log in for availability1 * 1 Items3.640,00 kr
Exhaust filters
Product Information
- Katalog #DescriptionTilgængelighedPrice
Replacement hose connectors
Product Information
- Katalog #ThreadLengthTubing int.ØMaterialHole ØDescriptionTilgængelighedPrice
- NPT ¹/₈"40 mm9,0 - 10,5 mmPP7,0 mmHose connector, straight, milky whiteLog in for availability1 * 1 Items113,00 kr
Adaptors for exhaust filter
Product Information
- Katalog #DescriptionTilgængelighedPrice
Adaptors for exhaust filter connection
Product Information
- Katalog #DescriptionTilgængelighedPrice
Replacement fittings
Product Information
- Katalog #DescriptionFor tubing ext.ØColourTilgængelighedPrice
Replacement fittings preparative
Product Information
- Katalog #DescriptionFor tubing ext.ØTilgængelighedPrice
Replacement blind fittings
Product Information
- Katalog #DescriptionTilgængelighedPrice