Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Methoxyflurane

Flammable
Irritant
Supplier:   Apollo Scientific
Methoxyflurane
Supplier:  Apollo Scientific
Certificates

2 Options Available Below

Product Details & Documents
Synonyms:  2,2-Dichloro-1,1-difluoroethyl methyl ether
  • CAS Number:76-38-0
  • Formula:C₃H₄Cl₂F₂O
  • MDL Number:MFCD00040144
  • Molecular Weight:164.961 g/mol
  • Boiling Pt:103…104.5 °C
  • Melting Pt:–35 °C
  • Density:1.426 g/cm³ (25 °C)
  • Flash Pt:37 °C
  • Storage Temperature:Ambient
  • UN:3271
  • ADR:3,III
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 1.040,00 kr
    1.040,00 kr
    1 * 5 g
       1.040,00 kr

  • Bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 4.190,00 kr
    4.190,00 kr
    1 * 25 g
       4.190,00 kr

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.