
Perfluoro(4-methylpent-2-ene) 90%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2070-70-4
:
Formula:C₆F₁₂
MDL Number:MFCD00153253
:300.05 g/mol
Boiling Pt:49 °C
Density:1.5873 g/cm³ (25 °C)
:GHS07
Storage Temperature:Ambient
Frequently Bought Together