Order Entry
Denmark
ContactUsLinkComponent
5-Fluoro-5'-deoxyuridine (Doxifluridine) 98%
5-Fluoro-5'-deoxyuridine (Doxifluridine) 98%
Supplier:  Apollo Scientific
Certificates
5-Fluoro-5'-deoxyuridine (Doxifluridine) 98%
Supplier:  Apollo Scientific

Specification Test Results

  • Appearance:
    White solid
  • Melting Point:
    189.6-190.4 degrees C
  • Purity:
    98%

Specification Test Results

Frequently Bought Together