Order Entry
Denmark
ContactUsLinkComponent
Lithium fluoride 99.99%
Toxic
Lithium fluoride 99.99%
Catalog # APOSPC4990R-100G
Supplier:  Apollo Scientific
CAS Number:  
Toxic
Lithium fluoride 99.99%
Catalog # APOSPC4990R-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :25.94 g/mol
Boiling Pt:1680 °C
Melting Pt:845 °C
Density:2.635 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:232-152-0
UN:3288
ADR:6.1,III
 :GHS06
CAS Number:7789-24-4
Formula:FLi
MDL Number:MFCD00011090