3-[3-(Trifluoromethyl)phenoxy]azetidine hydrochloride 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1188374-88-0
:
Formula:C10H11ClF3NO
MDL Number:MFCD05664837
:253.65 g/mol
Storage Temperature:Ambient
Frequently Bought Together