Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

2-Bromophenetole

Irritant
Supplier:   Apollo Scientific
2-Bromophenetole
Supplier:  Apollo Scientific
Certificates
2-Bromophenetole

3 Options Available Below

Product Details & Documents
Synonyms:  1-Bromo-2-ethoxybenzene, 2-Bromophenyl ethyl ether
  • CAS Number:583-19-7
  • Formula:C₈H₉BrO
  • MDL Number:MFCD00061092
  • Molecular Weight:201.06 g/mol
  • Boiling Pt:223 °C
  • Density:1.4223 g/cm³ (25 °C)
  • Storage Temperature:Ambient
  • EINECS:209-501-0
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    100 mg
    Log in for availability
    Log in for availability
    1 * 100 mg
    1 * 100 mg - kr165.00
    kr165.00
    1 * 100 mg
       kr165.00

  • Bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - kr220.00
    kr220.00
    1 * 1 g
       kr220.00

  • Bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - kr604.00
    kr604.00
    1 * 5 g
       kr604.00