Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Denmark
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Isopropylcyclohexanol
4-Isopropylcyclohexanol 95%
4-Isopropylcyclohexanol 95%
APOSOR937521-5G
:
Apollo Scientific
:
4-Isopropylcyclohexanol 95%
APOSOR937521-5G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 g
Seq:
3173
:
:
142.24 g/mol
:
Ambient
:
4621-04-9
:
C₉H₁₈O
:
MFCD00043476