Order Entry
Denmark
ContactUsLinkComponent
2-Methyl-2-hexanol 95%
2-Methyl-2-hexanol 95%
Catalog # APOSOR937436-25G
Supplier:  Apollo Scientific
CAS Number:  
2-Methyl-2-hexanol 95%
Catalog # APOSOR937436-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308
  • Environmentally Preferable:

Specifications

About this item

Synonyms: 1,1-Dimethyl-1-pentanol, 2-Methylhexan-2-ol
 :
 :116.2 g/mol
Storage Temperature:Ambient
CAS Number:625-23-0
Formula:C₇H₁₆O
MDL Number:MFCD00004486