Order Entry
Denmark
ContactUsLinkComponent
4-Isobutylcyclohexanecarboxylic acid (cis and trans mixture) 95%
4-Isobutylcyclohexanecarboxylic acid (cis and trans mixture) 95%
Catalog # APOSOR930868-5G
Supplier:  Apollo Scientific
CAS Number:  
4-Isobutylcyclohexanecarboxylic acid (cis and trans mixture) 95%
Catalog # APOSOR930868-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :184.28 g/mol
Storage Temperature:Ambient
CAS Number:38792-88-0
Formula:C₁₁H₂₀O₂