1-(Dimethylamino)-2-methylpentan-3-one 95%
Catalog # APOSOR906981-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:143.2267 g/mol
Storage Temperature:Ambient
CAS Number:51690-03-0
Formula:C₈H₁₇NO
MDL Number:MFCD13185946